Description
- Accommodates 6 to 7.7mm O.D. plain stem thermometer
- Sidearm with standard taper inner joint with drip tip
- Vertical tube has top outer and lower inner joint
- Supplied with open top cap and Viton™ O-ring
Specifications
Specifications
| Includes | Open top cap and Viton™ O-ring |
| Material | Borosilicate Glass (3.3) |
| Product Type | Distillation adapter |
| Standard Taper Joint No. | 19/22 |