Learn More
Synthware™ Distillation Receiver, Cow Type, with Hose Connection
GREENER_CHOICESDP
Catalog No. 31501030 Shop All Kemtech Products
Change view
Click to view available options
Standard Taper Joint:
14/20
19/22
24/40
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint
31-501-030 14/20
31-501-031 19/22
31-501-032 24/40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 31-501-030 Supplier Synthware™ Supplier No. D681420
Add to Cart
Add to Cart
With hose connection for application of vacuum
  • With three receivers spaced 40 deg. apart at 105 deg. angle between top outer joint and lower inner drip tip joints
  • All joints of same size
Specifications
Type Cow Type Distillation Receiver
Standard Taper Joint 14/20
WARNING: Cancer - www.P65Warnings.ca.gov