Learn More
Synthware™ Distilling Adapters, Bent, 105°
GREENER_CHOICESDP
Catalog No. 31500666 Shop All Kemtech Products
Change view
Click to view available options
Standard Taper Joint No.:
Top: 14/20, Bottom: 14/20
Top: 19/22, Bottom: 19/22
Top: 24/40, Bottom: 24/40
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint No.
31-500-666 Top: 14/20, Bottom: 14/20
31-500-667 Top: 19/22, Bottom: 19/22
31-500-668 Top: 24/40, Bottom: 24/40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 31-500-666 Supplier Synthware™ Supplier No. A891420
Add to Cart
Add to Cart
With an inner drip joint at the bottom and an outer joint at the top
Synthware distillation adapter with sealed in drip tip. Adapter is bent at 105 degrees. Standard taper top outer joint and lower inner joints.
Specifications
For Use With (Application) Organic Synthesis
Item Description Distilling Adapter, Bent, 105 Degree
Material Borosilicate Glass (3.3)
Product Type Distilling Adapter
Quantity 1
Standard Taper Joint No. Top: 14/20, Bottom: 14/20