Learn More
Synthware™ Drying Tube Adapters, Angled at 75°
GREENER_CHOICESDP
Catalog No. 31500653 Shop All Kemtech Products
Change view
Click to view available options
Standard Taper Joint No.:
14/20
19/22
24/40
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint No.
31-500-653 19/22
31-500-652 14/20
31-500-654 24/40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 31-500-653 Supplier Synthware™ Supplier No. A871922
Add to Cart
Add to Cart
Angled at 75° to avoid desiccant contact with reactions
Synthware drying tube angled at 75 degrees. Angled to avoid dessicant contact with reactions. With standard taper inner joint.
Specifications
For Use With (Application) Organic Synthesis
Item Description Drying Tube Adapter, Angled at 75 Degree
Material Borosilicate Glass (3.3)
Product Type Drying Tube Adapter
Quantity 1
Standard Taper Joint No. 19/22