Learn More
Synthware™ Drying Tube Adapters, Straight
GREENER_CHOICESDP
Catalog No. 31500649 Shop All Kemtech Products
Change view
Click to view available options
Standard Taper Joint No.:
14/20
19/22
24/40
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint No.
31-500-649 14/20
31-500-650 19/22
31-500-651 24/40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 31-500-649 Supplier Synthware™ Supplier No. A861420
Add to Cart
Add to Cart
With top standard taper outer joint and lower inner joint of same size
Synthware straight drying tube. With top outer and lower inner standard taper joints of the same size.
Specifications
For Use With (Application) Organic Synthesis
Material Borosilicate Glass (3.3)
Product Type Drying Tube Adapter
Quantity 1
Standard Taper Joint No. 14/20