Description
- With standard taper inner joints
- Available with either coarse or medium porosity fritted disc
Specifications
Specifications
| Material | Borosilicate Glass (3.3) |
| Sterility | Non-sterile |
| Porosity | Coarse |
| Standard Taper Joint | 14/20 |
| Diameter (Metric) | ∼20 mm O.D. |