Learn More
Synthware™ Filter Tubes with Fritted Disc
GREENER_CHOICESDP
Catalog No. 31501557 Shop All Kemtech Products
Change view
Click to view available options
Diameter (Metric):
∼20 mm O.D.
∼30 mm O.D.
∼36 mm O.D.
Porosity:
Coarse
Medium
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Porosity
31-501-557 ∼20 mm O.D. Medium
31-501-556 ∼20 mm O.D. Coarse
31-501-560 ∼36 mm O.D. Coarse
31-501-561 ∼36 mm O.D. Medium
31-501-558 ∼30 mm O.D. Coarse
31-501-559 ∼30 mm O.D. Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 31-501-557 Supplier Synthware™ Supplier No. F701420M
Add to Cart
Add to Cart
Used for filtration followed by recrystallization on opposite side of disc of air sensitive compounds
  • With standard taper inner joints
  • Available with either coarse or medium porosity fritted disc
Specifications
Material Borosilicate Glass (3.3)
Porosity Medium
Standard Taper Joint 14/20
Diameter (Metric) ∼20 mm O.D.