Description
- Standard taper inner joint at each end
- Available with either coarse or medium porosity fritted disc
- With two 2 mm glass stopcocks having constant pressure retention devices
Specifications
Specifications
| Description | Two neck pear shaped flask |
| Capacity (Metric) | 100 mL |
| Standard Taper Joint | Center: 14/20; Side: 14/20 |