Learn More
Synthware™ Flow Control Adapters with Glass Stopcock
GREENER_CHOICESDP
Catalog No. 31500566 Shop All Kemtech Products
Change view
Click to view available options
Standard Taper Joint No.:
14/20
19/22
24/40
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint No.
31-500-566 14/20
31-500-567 19/22
31-500-568 24/40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 31-500-566 Supplier Synthware™ Supplier No. A611420
Add to Cart
Add to Cart
To regulate the flow of gases or liquids into reaction systems
With 2mm bore stopcock
Specifications
Includes 2mm bore stopcock
Material Stopcock: Glass
Product Type Vacuum adapter
Standard Taper Joint No. 14/20