Learn More
Synthware™ Flow Control Adapters with Metering Stopcock, 90°
GREENER_CHOICESDP
Catalog No. 31500581 Shop All Kemtech Products
Change view
Click to view available options
Standard Taper Joint No.:
14/20
19/22
24/40
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint No.
31-500-581 14/20
31-500-582 19/22
31-500-583 24/40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 31-500-581 Supplier Synthware™ Supplier No. A721420
Add to Cart
Add to Cart
Allows delivery of preset volumes of gases or liquids to reactions using the needle valve
Provides on/off control with the main valve
Specifications
Material Borosilicate Glass (3.3)
Product Type Flow control adapter
Standard Taper Joint No. 14/20