Learn More
Synthware™ Flow Control Adapters with PTFE Stopcock and Outer Joint
GREENER_CHOICESDP
Catalog No. 31500560 Shop All Kemtech Products
Change view
Click to view available options
Standard Taper Joint No.:
14/20
19/22
24/40
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint No.
31-500-560 14/20
31-500-561 19/22
31-500-562 24/40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 31-500-560 Supplier Synthware™ Supplier No. A711420
Add to Cart
Add to Cart
For regulating the flow of gases or liquids into reaction systems
  • Stopcock has bore of 2mm
  • With standard taper inner joint
Specifications
Includes PTFE Stopcock
Material PTFE
Product Type Flow control adapter
Standard Taper Joint No. 14/20