Learn More
Synthware™ Flushig Adapters
GREENER_CHOICESDP
Catalog No. 31500585 Shop All Kemtech Products
Change view
Click to view available options
Standard Taper Joint No.:
14/20
24/40
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint No.
31-500-585 14/20
31-500-586 24/40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 31-500-585 Supplier Synthware™ Supplier No. A761420
Add to Cart
Add to Cart
For evacuation and transfer of product into another vessel with same size standard taper joints
T-bore PTFE stopcock has a 2mm bore
Specifications
Includes 2mm T-bore PTFE stopcock
Material PTFE
Product Type Flushig Adapter
Standard Taper Joint No. 14/20