Learn More
Synthware™ Flushing Adapters with PTFE Stopcock
GREENER_CHOICESDP
Catalog No. 31500571 Shop All Kemtech Products
Change view
Click to view available options
Standard Taper Joint No.:
14/20
19/22
24/40
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint No.
31-500-571 14/20
31-500-572 19/22
31-500-573 24/40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 31-500-571 Supplier Synthware™ Supplier No. A681420
Add to Cart
Add to Cart
Allows systems to be flushed with an inert gas while sampling directly through the bore of the T-bore stopcock
Synthware flushing adapter is used to flush and fill attached vessels with an inert gas to provide protective storage.
Specifications
For Use With (Application) Organic Synthesis
Item Description Flushing Adapter with PTFE Stopcock
Material Borosilicate Glass (3.3)
Product Type Flushing Adapter
Quantity 1
Standard Taper Joint No. 14/20