Specifications
| For Use With (Application) | Organic Synthesis |
| Item Description | Flushing Adapter with PTFE Stopcock |
| Material | Borosilicate Glass (3.3) |
| Product Type | Flushing Adapter |
| Quantity | 1 |
| Standard Taper Joint No. | 19/22 |