Learn More
Synthware™ Gas Dispersion Tubes, Fritted
GREENER_CHOICESDP
Catalog No. 31501544 Shop All Kemtech Products
Change view
Click to view available options
Porosity:
Coarse
Extra Coarse
Medium
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Porosity
31-501-544 Coarse
31-501-543 Extra Coarse
31-501-545 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 31-501-544 Supplier Synthware™ Supplier No. F260810C
Add to Cart
Add to Cart
For use with filtration techniques
  • With stem O.D. of 8 mm and Length: 150 mm
  • Disc diameter: 10 mm
Specifications
Porosity Coarse