Description
- Hose connection used for connection to vacuum source
- Stopcock allows control of vacuum
- With 2mm straight-bore PTFE or Glass stopcock
Specifications
Specifications
| Material | Stopcock: Glass |
| Product Type | Inlet adapter with stopcock and drip tip |
| Standard Taper Joint No. | 14/20 |