Learn More
Synthware™ Inlet Adapters
GREENER_CHOICESDP
Catalog No. 31500524 Shop All Kemtech Products
Change view
Click to view available options
Standard Taper Joint No.:
14/20
19/22
24/40
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint No.
31-500-524 19/22
31-500-523 14/20
31-500-525 24/40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 31-500-524 Supplier Synthware™ Supplier No. A451922
Add to Cart
Add to Cart
For use with gas inlet tubes or plain stem thermometers
Supplied with one holed rubber adapter
Specifications
Material Borosilicate Glass
Product Type Inlet Adapter
Standard Taper Joint No. 19/22