Learn More
Synthware™ Inlet or Thermometer Adapters
GREENER_CHOICESDP
Catalog No. 31500527 Shop All Kemtech Products
Change view
Click to view available options
Standard Taper Joint No.:
14/20
19/22
24/40
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint No.
31-500-527 19/22
31-500-526 14/20
31-500-528 24/40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 31-500-527 Supplier Synthware™ Supplier No. A521922
Add to Cart
Add to Cart
With a compression cap and Viton O-ring to form a vacuum tight seal
  • Cap accommodates 5.0 to 7.5mm O.D. thermometer
  • Supplied with one Nylon™ cap and one Viton™ O-ring
Specifications
Material Borosilicate Glass
Product Type Inlet or Thermometer Adapter
Standard Taper Joint No. 19/22