Learn More
Synthware™ Joint Clip, Standard Taper Joint
GREENER_CHOICESDP
Catalog No. 31502284 Shop All Kemtech Products
Change view
Click to view available options
Standard Taper Joint:
14/20
19/22
24/40
29/42
34/45
40/50
45/50
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint
31-502-284 40/50
31-501-921 14/20
31-501-922 19/22
31-501-923 24/40
31-501-924 29/42
31-501-925 34/45
31-501-926 45/50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 31-502-284 Supplier Synthware™ Supplier No. CS004050
Add to Cart
Add to Cart
Used to help hold together standard taper joints
  • Clips are made from polyacetal plastic
Specifications
For Use With (Equipment) Filtration Glassware
Material Polyacetal Plastic
Standard Taper Joint 40/50