Learn More
Synthware™ Kjeldahl Trap Adapter
GREENER_CHOICESDP
Catalog No. 31500700 Shop All Kemtech Products
Change view
Click to view available options
Standard Taper Joint:
24/40
Catalog No. Standard Taper Joint
31-500-700 24/40
1 options
Catalog No. 31-500-700 Supplier Synthware™ Supplier No. A172440
Add to Cart
Add to Cart
Features two standard taper inner joints parallel to one another with Kjeldahl trap at lower end
Distance between center of joints is approximate 200 mm
Specifications
Type Kjeldahl Trap Adapter
Standard Taper Joint 24/40