Learn More
Description
- Joints of same size
Specifications
Specifications
| Standard Taper Joint No | 24/40 |
| Material | Glass |
| Length (Metric) Jacket | 300 mm |
| Type | Liebig Condenser |
| Description | Hose connection: 8mm O.D. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

