Description
- With standard taper outer joint and side arm with high vacuum valve
- Valves with PTFE O-ring at tip and PTFE wiper to protect Viton O-ring on shaft
Specifications
Specifications
| Description | 50 mL, 0-4 mm |
| Cone Size | 14/20 |
| Product Type | Reaction Tube with High-vacuum Valve |