Learn More
Synthware™ Reducing Adapters
GREENER_CHOICESDP
Catalog No. 31500507 Shop All Kemtech Products
Change view
Click to view available options
Standard Taper Joint No.:
Top: 10/30, Bottom: 24/40
Top: 14/20, Bottom: 19/22
Top: 14/20, Bottom: 24/40
Top: 19/22, Bottom: 24/40
Top: 24/40, Bottom: 29/42
Top: 24/40, Bottom: 34/45
Top: 24/40, Bottom: 45/50
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint No.
31-500-507 Top: 14/20, Bottom: 19/22
31-500-506 Top: 10/30, Bottom: 24/40
31-500-508 Top: 14/20, Bottom: 24/40
31-500-509 Top: 19/22, Bottom: 24/40
31-500-510 Top: 24/40, Bottom: 29/42
31-500-511 Top: 24/40, Bottom: 34/45
31-500-512 Top: 24/40, Bottom: 45/50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 31-500-507 Supplier Synthware™ Supplier No. A201419
Add to Cart
Add to Cart
With standard taper joints at top and bottom
Synthware reducing adapters are used for connecting dissimilar standard taper joint sizes. The lower inner joint is larger in size than the top outer joint.
Specifications
For Use With (Application) Organic Synthesis
Material Borosilicate Glass (3.3)
Product Type Reducing adapter
Quantity 1
Standard Taper Joint No. Top: 14/20, Bottom: 19/22