Description
- Adapter placed between vapor tube and evaporation flask on rotary evoprator to prevent contents of flask to be drawn into condensor in event of bumping
- Sealed-in fritted disc reduces foam from being carried over
Specifications
Specifications
| Standard Taper Joint | Upper: 24/40, Lower: 14/20 |
| Material | Borosilicate Glass (3.3) |
| Product Type | Anti-splash Adapter with Fritted Disc |
| Capacity (Metric) | 250 mL |