Description
- Placed between vapor tube and evaporation flask on rotary evoprator
- Prevent contents of flask to be drawn into condensor in event of bumping
Specifications
Specifications
| Standard Taper Joint | Upper: 29/42, Lower: 14/20 |
| Product Type | Anti-splash Adapter |
| Material | Borosilicate Glass (3.3) |
| Capacity (Metric) | 100 mL |