Description
- Placed between vapor tube and evaporation flask on rotary evoprator
- Prevent contents of flask to be drawn into condensor in event of bumping
Specifications
Specifications
| Standard Taper Joint | Upper: 24/40, Lower: 19/22 |
| Material | Borosilicate Glass (3.3) |
| Capacity (Metric) | 250 mL |
| Product Type | Rotary Evaporator, Bump Trap, Adapter |