Description
- Threaded side port for insertion of thermometer or bleed tube having O.D. between 5 mm and 7.5 mm
- Heavy wall design
- All sizes hand blown from tubing to ensure uniform wall thickness
Specifications
Specifications
| Description | Joint: 14/20; With threaded inlet |
| Capacity (Metric) | 250 mL |