Description
- All sizes are hand blown from tubing to ensure uniform wall thickness
Specifications
Specifications
| Description | Round bottom flask, 4-Neck |
| Capacity (Metric) | 500 mL |
| Standard Taper Joint | Center: 24/40; 24/40 side |