Description
- High vacuum valve with PTFE O-ring at tip and PTFE wiper (“umbrella”) to protect Viton O-ring on shaft
Specifications
Specifications
| Description | Valve Size: 0-8mm |
| Diameter (Metric) Outer | 28 mm |
| Overall Length (Metric) | 120 mm |
| Product Type | Storage Tube with High-vacuum Valve |