Learn More
Synthware™ Vacuum/Inert Gas Adapters with T-bore Glass or PTFE Resin Stopcock
Shop All Kemtech Products
Description
- With 4mm glass or PTFE Resin stopcock bore
- Glass stopcocks have constant pressure retention device
Specifications
Specifications
| Material | Stopcock: PTFE Resin |
| Product Type | Vacuum/Inert Gas adapter |
| Standard Taper Joint No. | 14/20 |