Learn More
Synthware™ Vacuum Transfer Adapter with PTFE Stopcocks
GREENER_CHOICESDP
Catalog No. 31500589 Shop All Kemtech Products
Change view
Click to view available options
Standard Taper Joint No.:
14/20
19/22
24/40
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint No.
31-500-589 19/22
31-500-588 14/20
31-500-590 24/40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 31-500-589 Supplier Synthware™ Supplier No. A121922
Add to Cart
Add to Cart
Adapter with 2mm stopcock used for the transfer of sensitive and hazardous solutions
Synthware vacuum transfer adapter with PTFE stopcocks and standard taper inner joint. Used for the transfer of sensitive and hazardous solutions while under vacuum.
Specifications
For Use With (Application) Organic Synthesis
Material Borosilicate Glass (3.3)
Product Type Glass Joint Adapter
Quantity 1
Standard Taper Joint No. 19/22