Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Syntri Safeguard Tissue Processing Reagents

Catalog No. 6115SSS Shop All Epredia Products
Change view
Click to view available options
Product Type:
10% Neutral Buffered Formalin
Clear-Rite 3
Dehydrant 100%
Fixation Buffer
Flex 100
Formal-Fixx
Isopropyl Alcohol
Tissue Processing Reagent
Xylene
Xylene Substitute
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
61-15S-SS Fixation Buffer
6915SS Clear-Rite 3
6764515SS Formal-Fixx
6615SS Xylene
6215SS Dehydrant 100%
8115SS Flex 100
9517SS Isopropyl Alcohol
5735SS 10% Neutral Buffered Formalin
6764215SS Xylene Substitute
61-15S-S Tissue Processing Reagent
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 61-15S-SS Supplier Epredia™ Supplier No. 6115SSS

Tissue processing reagents that feature RFID technology for reagent management on the Epredia™ Revos™ Tissue Processor.

The Syntri Safeguard Reagents come in 5 liter bottles for tissue processing with RFID tags embedded in the labels. When partnered with the Epredia™ Revos™ Tissue Processor, Syntri Safeguard Reagent usage is tracked and logged on the instrument. The technology remembers every bottle loaded, documents every protocol executed and maintenance performed. Helping to reduce downtime and facilitates proper use of reagents.

Specifications

For Use With (Equipment) All Tissue Processors
Grade Histological
Product Type Fixation Buffer
Quantity 4 x 5 L
Solution Type Fixation Buffer
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.