Learn More
Medchemexpress LLC HY-135902A 5mg Medchemexpress, Synucleozid (hydrochloride) CAS:0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY135902A5MG
Encompass_Preferred
Medchemexpress, HY-135902A 5mg Synucleozid (hydrochloride) CAS:0 Synucleozid hydrochloride (NSC 377363 hydrochloride) is a potent inhibitor of the SNCA mRNA that encodes α-synuclein protein (IC50=1.5 μM). Synucleozid selectively targets the α-synuclein mRNA 5′ UTR at the designed IRE site, decreases the amount of SNCA mRNA loaded into polysomes and thereby inhibits SNCA translation. Synucleozid has the potential for the investigation of Parkinson’s disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0382
Due to product restrictions, please to purchase or view availability for this product.