Learn More
Chemglass Life Sciences Syringe Filter, 0.45µm PVDF Membrane, 25mm Housing Diameter, Individually Wrapped, Sterile
SDP
Supplier:  Chemglass Life Sciences CGN2005011
Encompass_Preferred
NEST Syringe Filters are high quality filters individually wrapped and sterile. The standard range of filters are available in PVDF (Polyvinylidene Fluoride) and PES (Polyethersulfone). All filters are designed with Female Luer-Lock inlets and Male Luer Slip outlets. Sterilized non-pyrogenic and RNA/DNA-Free. The syringe filter housings are polypropylene. PVDF ¿ Extremely low protein-binding; for filtration of non-aggressive aqueous and mild organic solutions or were maximizing protein recovery is important
Catalog No. 50-189-7733
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.