Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Syringaldazine 99+%

Catalog No. AC175930010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC175930010 1 g
AC175930050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC175930010 Supplier Thermo Scientific Chemicals Supplier No. 175930010

CAS: 14414-32-5 | C18H20N2O6 | 360.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14414-32-5
Molecular Formula C18H20N2O6
InChI Key MYNVJNZGTAVRJD-UHFFFAOYSA-N
Synonym syringaldazine, 4-hydroxy-3,5-dimethoxybenzaldehyde azine, 3,5-dimethoxy-4-hydroxybenzalazine, 4,4'-hydrazine-1,2-diylidenebis methanylylidene bis 2,6-dimethoxyphenol, benzaldehyde, 4-hydroxy-3,5-dimethoxy-, 4-hydroxy-3,5-dimethoxyphenyl methylene hydrazone, 4-1e-e-2-4-hydroxy-3,5-dimethoxyphenyl methylidene hydrazin-1-ylidene methyl-2,6-dimethoxyphenol, 4-2-4-hydroxy-3,5-dimethoxyphenyl methylidene hydrazin-1-ylidene methyl-2,6-dimethoxyphenol, benzaldehyde, 4-hydroxy-3,5-dimethoxy-, 2-4-hydroxy-3,5-dimethoxyphenyl methylene hydrazone
PubChem CID 5379425

Specifications

Melting Point 209°C to 213°C
Color Green to Yellow
Infrared Spectrum Authentic
Quantity 1 g
SMILES COC1=CC(=CNNC=C2C=C(C(=O)C(=C2)OC)OC)C=C(C1=O)OC
IUPAC Name 4-[[2-[(3,5-dimethoxy-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]hydrazinyl]methylidene]-2,6-dimethoxycyclohexa-2,5-dien-1-one
Molecular Weight (g/mol) 360.366
Formula Weight 360.36
Percent Purity ≥99.0%
Physical Form Fine Crystalline Powder
Chemical Name or Material Syringaldazine, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.