Learn More
Med Vet International Dynarex Suction Connection Tubing w/Male Connector, Non-Conductive, 3/16" x 6' Tube, 50/Case
Supplier:  Med Vet International 32101
Encompass_Preferred
Dynarex Resp-O2™ Suction Connecting Tubing is designed to provide secure connections to both portable and in-wall suction sources. This non-conductive Tubing features female connectors on both ends and includes an optional male (straw) adapter for additional attachments. Designed to connect suction sources to suction catheters and yankauers Constructed from transparent material for clear visualization during suctioning Durable, crush-resistant tubing Single-use and disposable Not made with natural rubber latex or DEHP
Catalog No. 50-304-4677
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.