Learn More
STEMCELL Technologies Syringe Filter, Nylon, 0.22 μm, 30 mm, Size: 30 Units
SDP
Supplier:  STEMCELL Technologies 2000589
Encompass_Preferred
Remove fine particles and unwanted materials with syringe filters. The nylon membrane of these filters is ideal for universal analytical procedures and chemical filtration. Each individually wrapped syringe filter is sterile non-pyrogenic and RNase/DNase-free and features a housing made with virgin polypropylene. They have a female luer lock inlet and a male luer slip outlet.
Catalog No. 50-239-0177
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.