Learn More
Synthware™ Syringe Needles
GREENER_CHOICESDP
Catalog No. 31501990 Shop All Kemtech Products
Change view
Click to view available options
Length (Metric):
180 mm
200 mm
300 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
31-501-990 300 mm
31-501-866 200 mm
31-501-988 180 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 31-501-990 Supplier Synthware™ Supplier No. S380230
Add to Cart
Add to Cart
Long needle used for solvent retrieval.
Specifications
Material Stainless Steel
Needle Gauge 16 G
For Use With (Application) Organic Synthesis
Length (Metric) 300 mm
Product Type Stainless Steel Needle
Quantity 1 Ea.
For Use With (Equipment) Gas Chromatography Systems
Connection Type Luer Lock
Graduated No
Sterility Non-sterile