Learn More
MilliporeSigma™ Supelco™ Syringe for On-Column Injection
Catalog No. 111010891 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With (Equipment):
Agilent/HP Injectors
Catalog No. For Use With (Equipment)
11-101-0891 Agilent/HP Injectors
1 options
Catalog No. 11-101-0891 Supplier MilliporeSigma™ Supelco™ Supplier No. 21352
In this system, a 32-gauge needle is inserted in a cool, narrow bore injection valve and then into capillary column
TRUSTED_SUSTAINABILITY
Specifications
Material Stainless Steel (Tip)
Needle Gauge 32 G
Diameter (Metric) Outer Needle 0.24 mm
For Use With (Application) On-Column Injection
Product Type Syringe
Needle Point Number 3
Quantity 1 Ea.
Volume (Metric) 10 μL
Length (Metric) Needle 12.5 cm
Needle Point Style Blunt
For Use With (Equipment) Agilent/HP Injectors
Connection Type Fixed