Learn More
Med Vet International Nipro Syringe, 20mL, Luer Slip, 50/Box, JD+20ES
Supplier:  Med Vet International JD20ES
Encompass_Preferred
Nipro JD+20ES 20mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer Slip tip of this 20mL syringe fits conventional and safety needles. This quantity is 50/BX
Catalog No. 50-209-2982
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.