Learn More
Med Vet International SYRINGE3CC 20 X 1 1/2
Supplier:  Med Vet International 03L2038
Encompass_Preferred
Terumo SS-03L2038 3mL Syringe & Needle Combo comes with a clear barrel and easy to read graduation marks. The Luer Lock tip of this 3mL syringe fits conventional and safety needles. This Terumo SS-03L2038 3mL Syringe & Needle Combo comes with a 20GX 1 1/2" Thin Wall needle. Terumo's thin wall needles areprecisely honed and beveled. This makes for ease of penetration and less patient discomfort This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX
Catalog No. 50-255-9767
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.