Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Syringes Fitted with Luer Lock Valve

Catalog No. 111010420 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Volume (Metric):
1 mL
5 mL
10 mL
25 mL
100 mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
11-101-0420 1 mL
11-101-0219 5 mL
11-101-0152 10 mL
11-100-9457 25 mL
11-100-8589 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 11-101-0420 Supplier MilliporeSigma™ Supelco™ Supplier No. 509590
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Feature an interference fit between the plunger tip and the barrel to improve the syringe's gas sealing properties

  • Maximum inertness for liquid or gas samples
  • Suitable for manual dispenser/diluter applications
  • High accuracy of dispensed volume
  • Syringes are autoclavable when plunger is removed
TRUSTED_SUSTAINABILITY

Specifications

Accuracy ± 1%
Autoclavable Autoclavable
Material Borosilicate Glass (Barrel), PTFE (Plunger)
Description Pre-fitted with Luer Lock Valve
Product Type Syringe
Model 1 MDR-VLL-GT
Quantity 1 Ea.
Volume (Metric) 1 mL
Connection Type Luer Lock, Kel-F Luer Cones
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.