Learn More
MilliporeSigma™ Supelco™ Syringes with Removable Needle Valve
Catalog No. 111010301 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Volume (Metric):
100 μL
250 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
11-101-0301 100 μL
11-101-0128 250 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-0301 Supplier MilliporeSigma™ Supelco™ Supplier No. 509531
Add to Cart
Add to Cart
Feature an interference fit between the plunger tip and the barrel, which enhances the gas sealing properties of the syringe
  • Maximum inertness for liquid or gas samples
  • Suitable for manual dispenser/diluter applications
  • High accuracy of dispensed volume
  • Syringes are autoclavable when plunger is removed
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Material Borosilicate Glass (Barrel), PTFE (Plunger), Stainless Steel (Tip)
Description Needle Valve in Hub
Needle Gauge 23 G
Diameter (Metric) Outer Needle 0.63 mm
Product Type Syringe
Model 100 R-V-GT
Needle Point Number 2
Quantity 1 Ea.
Volume (Metric) 100 μL
Length (Metric) Needle 50 mm
Needle Point Style Bevel
Connection Type Kel-F Luer Cones, Removable Needle Valve