Learn More
MilliporeSigma™ Supelco™ Syringes for Shimadzu™ Autosamplers
Catalog No. 111010703 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Model:
10 R-S-0.47
10 R-S-0.63
10 R-S-GT-0.63
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
11-101-0703 10 R-S-0.47
11-101-0704 10 R-S-0.63
11-101-0812 10 R-S-GT-0.63
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-101-0703 Supplier MilliporeSigma™ Supelco™ Supplier No. 24418
Add to Cart
Add to Cart
For use with Shimadzu™ AOC14, 17, 20 and 20i Autosamplers
TRUSTED_SUSTAINABILITY
Specifications
Material Stainless Steel (Tip)
Description Dark Orange Patch on Barrel for Easy Volume Identification
Needle Gauge 26 G
Product Type Syringe
Model 10 R-S-0.47
Needle Point Number 1
Quantity 1 Ea.
Volume (Metric) 10 μL
Length (Metric) Needle 42 mm
Needle Point Style Cone
For Use With (Equipment) Shimadzu AOC 14, 17, 20 and 20i GCs
Show More Show Less
Shimadzu is a trademark of Shimadzu Corporation