Learn More
Apexbio Technology LLC Syringic acid 530-57-4 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC N22975.1
Encompass_Preferred
Syringic acid (CAS 530-57-4) is a small-molecule inhibitor targeting low-density lipoprotein (LDL) oxidation It is designed to inhibit LDL oxidation thereby modulating oxidative stress pathways Syringic acid exerts its biological activity primarily through reactive oxygen species scavenging In in vitro studies syringic acid demonstrates inhibitory activity with an IC50 value of approximately 5 8 M in copper-induced LDL oxidation assays Based on these pharmacological properties syringic acid holds research potential in studies related to oxidative stress atherosclerosis progression and cardiovascular disease models
Catalog No. 50-000-96515