Learn More
TARGETMOL CHEMICALS INC SYROSINGOPINE 5MG
Supplier: TARGETMOL CHEMICALS INC TN2252
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Syrosingopine has selective depleting effect on brain amines is potentiated by combined treatment with disulfiram or fusaric acid a dopamine beta-hydroxylase inhibitor. purity: 99%
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.