Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ System Reagents for Phadia™ 1000

Catalog No. 10943901P
Change view
Click to view available options
For Use With (Equipment):
Phadia 1000
Phadia™ 1000
Quantity:
∼1200
2 Procedures
2 x 5 L
6 x ∼1200
Product Type:
Assay System Reagent for Phadia 1000
Maintenance solution kit
Stop solution
Washing solution
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Quantity Product Type
10-943-901P Phadia 1000 6 x ∼1200 Assay System Reagent for Phadia 1000
34-227-151P Phadia 1000 ∼1200 Stop solution
10-920-201P Phadia™ 1000 2 x 5 L Washing solution
10-947-601P Phadia™ 1000 2 Procedures Maintenance solution kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 10-943-901P Supplier Thermo Scientific™ Supplier No. 10943901
Add to Cart
Add to Cart
This item is not returnable. View return policy

Replacement reagents provide true accuracy, high precision and outstanding quality

TRUSTED_SUSTAINABILITY

Specifications

Product Type Assay System Reagent for Phadia 1000
For Use With (Equipment) Phadia 1000
Quantity 6 x ∼1200
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.