Learn More
Cytiva Ultralab™ Systems with 115 V pump - 5 L Ultralab™ system (consists of an Ultrareservoir container, MasterFlex L/S variable speed peristaltic pump, and Ultrasette accessory kit; connects to Ultrasette device sold separately) (1/pkg)
Supplier:  Cytiva FS007X70
Encompass_Preferred
Simplify Processing of 100 mL to 5 L Volumes When Using the Ultrasette™ Device-Self-contained siphoning units allow continuous buffer exchange-Uses less liquid with less handling and mess than conventional dialysis systems-Constructed of clear acrylic with easy-to-read volume graduations-Connections located in base plate for easy access and minimal sample hold-up-Feed pressure gauge monitors sample processing-Three-way sampling valve allows maximum sample recovery without decanting-Peristaltic pump action minimizes shear of sensitive molecules, eliminates air in the system, and reduces foaming-A variable speed controller allows precise adjustment of crossflow during sample processing This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3462
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.