Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SYT17 Polyclonal Antibody, Invitrogen™

Catalog No. PA550271
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550271 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550271 Supplier Thermo Scientific Supplier No. PA550271

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total SYT17 protein.

Plays a role in dendrite formation by melanocytes. Classified as a brain and kidney protein (not to be confused with the BigK K+ channel); may play a role in cAMP mediated signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SYT17
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene SYT17
Gene Accession No. Q920M7, Q9BSW7
Gene Alias B/K protein; Bk; brain and kidney protein; protein B/K; synaptotagmin 17; synaptotagmin XVII; Synaptotagmin-17; Syt17; SytXVII
Gene Symbols SYT17
Host Species Rabbit
Immunogen Fusion protein corresponding to residues near the N terminal of human synaptotagmin XVII
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110058, 51760
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.