Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SYT4 Mouse anti-Human, Clone: UMAB128, lyophilized, UltraMAB™
SDP

Catalog No. 501685010
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-5010 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-5010 Supplier OriGene Supplier No. UM500093CF
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

May be involved in Ca(2+)-dependent exocytosis of secretory vesicles through Ca(2+) and phospholipid binding to the C2 domain or may serve as Ca(2+) sensors in the process of vesicular trafficking and exocytosis.

Specifications

Antigen SYT4
Applications Immunohistochemistry (Paraffin), Peptide Array
Classification Monoclonal
Clone UMAB128
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative
Gene SYT4
Gene Accession No. Q9H2B2
Gene Alias HsT1192
Gene Symbols SYT4
Host Species Mouse
Immunogen Full length human recombit protein of human SYT4 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6860
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.